siRNA Details
Gene Symbol: Mks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039684.2 | NP_001034773.2 | 7, 8 | 772 | ||
GenBank mRNA | AK030982.1 | - | 7, 8 | 752 | ||
AK138267.1 | - | 6, 7 | 742 | |||
AK170482.1 | - | 7, 8 | 772 | |||
BC150995.1 | - | 7, 8 | 840 | 9087908 | ||
BC158122.1 | - | 7, 8 | 932 | 9088195 | ||
DQ177342.1 | - | 7, 8 | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
