siRNA Details
Gene Symbol: Lsmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033437.2 | NP_001028609.1 | 3, 4 | 373 | ||
GenBank mRNA | AK142184.1 | - | 3, 4 | 374 | ||
BC147386.1 | - | 3, 4 | 293 | 9056448 | ||
BC147387.1 | - | 3, 4 | 293 | 9056444 | ||
BC172052.1 | - | 3, 4 | 297 |
