siRNA Details
Gene Symbol: Lsmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033437.2 | NP_001028609.1 | 2 | 227 | ||
GenBank mRNA | AK134025.1 | - | 403 | |||
AK142184.1 | - | 2 | 228 | |||
BC147386.1 | - | 2 | 147 | 9056448 | ||
BC147387.1 | - | 2 | 147 | 9056444 | ||
BC172052.1 | - | 2 | 147 |
