siRNA Details
Gene Symbol: Serpina11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166350.1 | NP_001159822.1 | 3 | 822 | ||
NM_199314.2 | NP_955018.2 | 3 | 836 | |||
GenBank mRNA | AK149420.1 | - | 3 | 800 | ||
BC024087.1 | - | 3 | 814 | 5101271 | ||
BC096459.1 | - | 4 | 728 | |||
BC096585.1 | - | 3 | 783 | 5097495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
