siRNA Details
Gene Symbol: Serpinb1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173051.2 | NP_766639.2 | 7 | 812 | ||
GenBank mRNA | BC104332.2 | - | 6 | 795 | 40045807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
