siRNA Details
Gene Symbol: Lyrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201358.2 | NP_958746.1 | 2 | 318 | ||
GenBank mRNA | AK050553.1 | - | 2 | 293 | ||
BC034664.1 | - | 2 | 216 | 5355517 | ||
BC080772.1 | - | 2 | 193 | 30139193 |

More Information
Other Species Targeted

Gene Ontology Categories:
