siRNA Details
Gene Symbol: Lyrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201358.2 | NP_958746.1 | 2 | 345 | ||
GenBank mRNA | AK050553.1 | - | 2 | 320 | ||
BC034664.1 | - | 2 | 243 | 5355517 | ||
BC080772.1 | - | 2 | 220 | 30139193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
