siRNA Details
Gene Symbol: Gm904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033770.1 | NP_001028942.1 | 2, 3 | 289 | ||
GenBank mRNA | AK131725.1 | - | 2, 3 | 289 | ||
BC147620.1 | - | 2, 3 | 279 | 9056610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
