siRNA Details
Gene Symbol: Zfp729b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163246.2 | NP_001156718.1 | 4 | 443 | ||
GenBank mRNA | AK137425.1 | - | 4 | 444 | ||
AK170565.1 | - | 2 | 159 | |||
BB629152.1 | - | 4 | 444 | |||
BC047145.1 | - | 4 | 399 | 6467898 |
