siRNA Details
Gene Symbol: Lrch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033439.3 | NP_001028611.2 | 4 | 847 | ||
NM_001252132.1 | NP_001239061.1 | 4 | 847 | |||
GenBank mRNA | AK129264.1 | - | 4 | 836 | ||
AK136026.1 | - | 4 | 453 | |||
AK143909.1 | - | 4 | 846 | |||
AK160902.1 | - | 2 | 223 | |||
AK161894.1 | - | 4 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
