siRNA Details
Gene Symbol: Dgkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081336.1 | NP_001074805.1 | 23 | 2707 | ||
NM_001253766.1 | NP_001240695.1 | 21 | 2395 | |||
NM_001281794.1 | NP_001268723.1 | 21 | 2395 | |||
GenBank mRNA | AK133314.1 | - | 22 | 3170 | ||
BC117904.1 | - | 21 | 2379 | |||
BC127173.1 | - | 21 | 2379 | 40097093 | ||
CK780920.1 | - | 3 | 230 |
