siRNA Details
Gene Symbol: Dgkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081336.1 | NP_001074805.1 | 21 | 2515 | ||
NM_001253766.1 | NP_001240695.1 | 19 | 2203 | |||
NM_001281794.1 | NP_001268723.1 | 19 | 2203 | |||
GenBank mRNA | AK083819.1 | - | 19 | 2204 | ||
AK133314.1 | - | 20 | 2978 | |||
BC117904.1 | - | 19 | 2187 | |||
BC127173.1 | - | 19 | 2187 | 40097093 | ||
CK780920.1 | - | 1 | 37 |
