siRNA Details
Gene Symbol: Higd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002900.1 | NP_001002900.1 | 3 | 179 | ||
GenBank mRNA | AB099517.1 | - | 3 | 179 | ||
AB110950.1 | - | 3 | 693 | |||
AB253942.1 | - | 3 | 219 |
Gene Symbol: Methig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024672.3 | NP_001019843.1 | 3 | 642 | ||
GenBank mRNA | AB099516.1 | - | 3 | 679 | ||
AK144032.1 | - | 3 | 643 | |||
BC139180.1 | - | 3 | 676 | 8862202 | ||
BC139182.1 | - | 3 | 676 | 8862204 | ||
BC145570.1 | - | 2 | 579 |
