siRNA Details
Gene Symbol: Ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159421.1 | NP_001152893.1 | 3 | 641 | ||
NM_001159422.1 | NP_001152894.1 | 4 | 418 | |||
NM_198645.2 | NP_941047.1 | 4 | 767 | |||
GenBank mRNA | AK135450.1 | - | 4 | 347 | ||
AV583692.1 | - | 4 | 418 | |||
BB686073.1 | - | 2 | 82 | |||
BC024406.1 | - | 4 | 331 | |||
BC024862.1 | - | 4 | 742 | 5356370 | ||
BC053690.1 | - | 4 | 319 | |||
CF617453.1 | - | 4 | 745 | |||
CJ094333.1 | - | 3 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
