siRNA Details
Gene Symbol: Gm1604b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033442.3 | NP_001028614.1 | 4 | 496 | ||
GenBank mRNA | AK162826.1 | - | 4 | 473 | ||
BC147040.1 | - | 4 | 289 | 9056052 | ||
BC147041.1 | - | 4 | 289 | 9056032 |
