siRNA Details
Gene Symbol: Zfp229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164676.1 | NP_001158148.1 | 5 | 566 | ||
GenBank mRNA | AK139846.1 | - | 5 | 356 | ||
AK147958.1 | - | 566 | ||||
BC043476.1 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
