siRNA Details
Gene Symbol: Ccdc78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165929.1 | NP_001159401.1 | 2 | 234 | ||
GenBank mRNA | AK140238.1 | - | 2 | 235 |

More Information
Other Species Targeted

Gene Ontology Categories:
