siRNA Details
Gene Symbol: H2-Eb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033978.3 | NP_001029150.1 | 2 | 319 | ||
GenBank mRNA | AK137912.1 | - | 2 | 320 | ||
AK157199.1 | - | 2 | 330 | |||
BC150834.1 | - | 2 | 320 | 9087745 | ||
KJ650234.1 | - | 2 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
