siRNA Details
Gene Symbol: Arhgef33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145452.1 | NP_001138924.1 | 11 | 1310 | ||
GenBank mRNA | AK053944.1 | - | 7 | 944 |

More Information
Other Species Targeted

Gene Ontology Categories:
