siRNA Details
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033449.1 | NP_001028621.1 | 12, 13 | 1257 | ||
NM_001037812.2 | NP_001032901.1 | 12, 13 | 1216 | |||
GenBank mRNA | AK133112.1 | - | 13, 14 | 1241 | ||
AK161697.1 | - | 12, 13 | 1261 | |||
AK167733.1 | - | 12, 13 | 1218 | |||
AK170118.1 | - | 12, 13 | 1199 | |||
BC147243.1 | - | 12, 13 | 1236 | 9056304 | ||
BC147244.1 | - | 12, 13 | 1236 | 9056305 |

More Information
Other Species Targeted

Gene Ontology Categories:
