siRNA Details
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 4 | 863 | ||
NM_001301745.1 | NP_001288674.1 | 4 | 857 | |||
GenBank mRNA | AK152278.1 | - | 4 | 731 | ||
AK152621.1 | - | 3 | 397 | |||
AK152683.1 | - | 4 | 731 | |||
AK152786.1 | - | 4 | 740 | |||
AK166217.1 | - | 4 | 863 | |||
BC132314.1 | - | 4 | 684 | 40130591 | ||
BC132316.1 | - | 4 | 684 | 40130593 | ||
BC145397.1 | - | 4 | 678 | 9053955 | ||
M74123.1 | - | 4 | 718 | |||
S62227.1 | - | 4 | 718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
