siRNA Details
Gene Symbol: 6330403A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081227.2 | NP_001074696.1 | 1 | 353 | ||
GenBank mRNA | AK031805.1 | - | 1 | 317 | ||
BC120654.1 | - | 1 | 208 | 40129577 | ||
BC120656.1 | - | 1 | 208 | 40129579 | ||
DN174476.1 | - | 1 | 353 |
