siRNA Details
Gene Symbol: 4921509C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198655.2 | NP_941057.1 | 2 | 268 | ||
GenBank mRNA | AK029504.1 | - | 2 | 270 | ||
BC119071.1 | - | 1 | 98 | 8733820 | ||
BC119073.1 | - | 1 | 98 | 8733822 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
