siRNA Details
Gene Symbol: Zfp663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005425.1 | NP_001005425.1 | 4 | 464 | ||
GenBank mRNA | AK141109.1 | - | 5 | 604 | ||
BC067000.1 | - | 4 | 464 | 5705933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
