siRNA Details
Gene Symbol: 2810408M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007581.1 | NP_001007582.1 | 2 | 1384 | ||
GenBank mRNA | AK013049.1 | - | 2 | 1394 | ||
AK077144.1 | - | 2 | 1393 | |||
AK167662.1 | - | 2 | 1392 | |||
BC085263.1 | - | 2 | 1384 | 6395509 |
