siRNA Details
Gene Symbol: Gm5148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198657.2 | NP_941059.1 | 3 | 878 | ||
GenBank mRNA | AK039231.1 | - | 3 | 954 | ||
AK039519.1 | - | 3 | 879 | |||
AK039710.1 | - | 3 | 879 | |||
AK045832.1 | - | 2 | 898 | |||
BC147192.1 | - | 2 | 675 | 9056236 | ||
BC147193.1 | - | 2 | 675 | 9056230 |
