siRNA Details
Gene Symbol: AI427809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144542.1 | - | 2 | 1044 | ||
AK171862.1 | - | 2 | 1061 | |||
BC147516.1 | - | 1 | 201 | 9056508 | ||
BC147547.1 | - | 1 | 201 | 9056538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
