siRNA Details
Gene Symbol: Gm13084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005371.3 | NP_001005371.2 | 2 | 307 | ||
GenBank mRNA | AK145369.1 | - | 2 | 304 | ||
BC080785.1 | - | 2 | 305 | 30481291 | ||
CJ082474.1 | - | 2 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
