siRNA Details
Gene Symbol: Tmem240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101506.1 | NP_001094976.1 | 3 | 518 | ||
GenBank mRNA | AK147532.1 | - | 2 | 4577 | ||
BC147820.1 | - | 4 | 379 | 9087537 | ||
BC147826.1 | - | 4 | 379 | 9087543 | ||
BY725301.1 | - | 3 | 450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
