siRNA Details
Gene Symbol: Lrcol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033459.3 | NP_001028631.1 | 2 | 262 | ||
GenBank mRNA | AK141249.1 | - | 2 | 263 | ||
BC147447.1 | - | 2 | 200 | 9056089 | ||
BC147469.1 | - | 2 | 200 | 9056111 | ||
BC171993.1 | - | 2 | 178 | |||
JQ258939.1 | - | 4 | 407 | |||
JQ258940.1 | - | 5 | 470 | |||
JQ258941.1 | - | 4 | 667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
