siRNA Details
Gene Symbol: Nxpe5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013773.3 | NP_001013795.3 | 6 | 2031 | ||
GenBank mRNA | AK156114.1 | - | 7 | 2100 | ||
BC055004.1 | - | 1647 | 6478735 | |||
BC106195.1 | - | 1829 | 6396133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
