siRNA Details
Gene Symbol: Tsen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199033.1 | NP_950198.1 | 9, 10 | 1170 | ||
GenBank mRNA | AK134697.1 | - | 9, 10 | 1166 | ||
BC061473.1 | - | 9, 10 | 1170 | 6414790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
