siRNA Details
Gene Symbol: Cracr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033464.3 | NP_001028636.1 | 5, 6 | 812 | ||
GenBank mRNA | AK143832.1 | - | 5, 6 | 813 | ||
BC150869.1 | - | 5, 6 | 890 | 9087780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
