siRNA Details
Gene Symbol: RAP1GDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100426.1 | NP_001093896.1 | 2, 3 | 303 | ||
NM_001100427.1 | NP_001093897.1 | 2, 3 | 300 | |||
NM_001100428.1 | NP_001093898.1 | 2, 3 | 303 | |||
NM_001100429.1 | NP_001093899.1 | 2, 3 | 300 | |||
NM_001100430.1 | NP_001093900.1 | 2, 3 | 303 | |||
NM_021159.4 | NP_066982.3 | 2, 3 | 303 | |||
GenBank mRNA | AF215923.1 | - | 1, 2 | 110 | ||
AF237413.1 | - | 1, 2 | 110 | |||
AK055032.1 | - | 2, 3 | 223 | |||
AK295156.1 | - | 2, 3 | 262 | |||
AY376900.1 | - | 1, 2 | 113 | |||
BC001816.2 | - | 2, 3 | 210 | 3010166 | ||
BC001851.2 | - | 2, 3 | 210 | 3010166 | ||
BC098269.1 | - | 2, 3 | 161 | 40001059 | ||
BC098334.1 | - | 2, 3 | 158 | 40001057 | ||
BC099708.1 | - | 2, 3 | 158 | 40001058 | ||
BC099845.1 | - | 2, 3 | 161 | 40001061 | ||
BT006837.1 | - | 1, 2 | 113 | |||
DC367998.1 | - | 2, 3 | 300 | |||
X63465.1 | - | 1, 2 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
