siRNA Details
Gene Symbol: RAP1GDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100426.1 | NP_001093896.1 | 9 | 1161 | ||
NM_001100427.1 | NP_001093897.1 | 9 | 1158 | |||
NM_001100428.1 | NP_001093898.1 | 8 | 1014 | |||
NM_001100429.1 | NP_001093899.1 | 8 | 1011 | |||
NM_001100430.1 | NP_001093900.1 | 7 | 885 | |||
NM_021159.4 | NP_066982.3 | 9 | 1161 | |||
GenBank mRNA | AF215923.1 | - | 8 | 968 | ||
AF237413.1 | - | 8 | 968 | |||
AK055032.1 | - | 9 | 1081 | |||
AK295156.1 | - | 8 | 973 | |||
AK297543.1 | - | 6 | 708 | |||
AK311243.1 | - | 6 | 696 | |||
AY376900.1 | - | 6 | 695 | |||
BC001816.2 | - | 9 | 1068 | 3010166 | ||
BC001851.2 | - | 9 | 1068 | 3010166 | ||
BC098269.1 | - | 9 | 1019 | 40001059 | ||
BC098334.1 | - | 8 | 869 | 40001057 | ||
BC099708.1 | - | 8 | 869 | 40001058 | ||
BC099845.1 | - | 8 | 872 | 40001061 | ||
BT006837.1 | - | 8 | 971 | |||
X63465.1 | - | 7 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
