siRNA Details
Gene Symbol: RAP1GDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100426.1 | NP_001093896.1 | 5, 6 | 699 | ||
NM_001100427.1 | NP_001093897.1 | 5, 6 | 696 | |||
NM_021159.4 | NP_066982.3 | 5, 6 | 699 | |||
GenBank mRNA | AF215923.1 | - | 4, 5 | 506 | ||
AF237413.1 | - | 4, 5 | 506 | |||
AK055032.1 | - | 5, 6 | 619 | |||
BC001816.2 | - | 5, 6 | 606 | 3010166 | ||
BC001851.2 | - | 5, 6 | 606 | 3010166 | ||
BC098269.1 | - | 5, 6 | 557 | 40001059 | ||
BT006837.1 | - | 4, 5 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
