siRNA Details
Gene Symbol: Prpmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024705.2 | NP_001019876.2 | 3 | 992 | ||
GenBank mRNA | AK132583.1 | - | 4 | 622 | ||
BC055856.1 | - | 3 | 983 | 4910526 | ||
BC061494.1 | - | 3 | 983 | |||
BC080748.1 | - | 3 | 983 | |||
BC092075.1 | - | 3 | 987 | 4920522 | ||
BC110664.1 | - | 3 | 990 | 4191067 | ||
X63004.1 | - | 881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
