siRNA Details
Gene Symbol: Rnf225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029809.2 | NP_084085.1 | 2 | 1010 | ||
GenBank mRNA | AK009344.1 | - | 2 | 1008 | ||
AK028523.1 | - | 2 | 1010 | |||
BC141220.1 | - | 2 | 1005 | 9055832 |
