siRNA Details
Gene Symbol: Rnf225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029809.2 | NP_084085.1 | 2 | 999 | ||
GenBank mRNA | AK009344.1 | - | 2 | 997 | ||
AK028523.1 | - | 2 | 999 | |||
BC141220.1 | - | 2 | 994 | 9055832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
