siRNA Details
Gene Symbol: Ppapdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080963.1 | NP_001074432.1 | 4 | 311 | ||
GenBank mRNA | AK081292.1 | - | 4 | 376 | ||
BC120497.1 | - | 4 | 311 | 40129420 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191631.
Gene Ontology Categories:
