siRNA Details
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 14 | 2510 | ||
GenBank mRNA | AK173222.1 | - | 12 | 3498 | ||
BC027735.1 | - | 14 | 2412 | |||
BC027790.1 | - | 7 | 1984 | |||
BC042391.1 | - | 7 | 2705 | |||
BC053460.1 | - | 2 | 549 | |||
BC059855.1 | - | 5 | 1043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
