siRNA Details
Gene Symbol: Cnep1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029074.3 | NP_083350.2 | 2 | 128 | ||
GenBank mRNA | AK017198.1 | - | 2 | 89 | ||
AK146578.1 | - | 2 | 119 | |||
BC018498.1 | - | 2 | 100 | |||
BC094385.1 | - | 2 | 87 | |||
BC118981.1 | - | 2 | 89 | |||
BC148291.2 | - | 2 | 89 | 40103934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
