siRNA Details
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145896.1 | NP_001139368.1 | 14 | 3523 | ||
NM_001145897.1 | NP_001139369.1 | 14 | 3467 | |||
NM_198671.2 | NP_941073.2 | 14 | 3655 | |||
GenBank mRNA | AK033263.1 | - | 14 | 3470 | ||
AK042915.1 | - | 14 | 3472 | |||
AK122216.1 | - | 10 | 2697 | |||
AK136004.1 | - | 4 | 966 | |||
AK154829.1 | - | 14 | 3502 | |||
BC043094.1 | - | 14 | 3655 | 6405464 |
