siRNA Details
Gene Symbol: Pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007462.1 | NP_001007463.1 | 5 | 634 | ||
GenBank mRNA | BC141316.1 | - | 5 | 633 | 9055928 | |
BC141317.1 | - | 5 | 633 | 9055929 | ||
BC145634.1 | - | 5 | 678 | 9054202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
