siRNA Details
Gene Symbol: Defa22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207658.4 | NP_997541.2 | 2 | 264 | ||
GenBank mRNA | AK008589.1 | - | 2 | 265 | ||
AY746427.1 | - | 2 | 221 |
Gene Symbol: Defa21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183253.3 | NP_899076.1 | 2 | 264 | ||
GenBank mRNA | AK008266.1 | - | 2 | 265 | ||
AY746426.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
