siRNA Details
Gene Symbol: Ccdc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166282.1 | NP_001159754.1 | 11 | 1816 | ||
NM_029212.3 | NP_083488.1 | 11 | 1465 | |||
GenBank mRNA | AB048860.1 | - | 11 | 1539 | ||
AK145264.1 | - | 11 | 1818 | |||
BC116291.1 | - | 11 | 1465 | 40094912 | ||
BC116292.1 | - | 11 | 1465 | 40094918 |
