siRNA Details
Gene Symbol: Ripply2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037907.1 | NP_001032996.1 | 4 | 260 | ||
GenBank mRNA | AB212223.1 | - | 4 | 260 | ||
AK081203.1 | - | 3 | 402 |

More Information
Other Species Targeted

Gene Ontology Categories:
