siRNA Details
Gene Symbol: Gm1123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080776.1 | NP_001074245.1 | 7 | 1055 | ||
GenBank mRNA | AB262662.1 | - | 7 | 1084 | ||
BC114986.1 | - | 7 | 1055 | 40056015 | ||
BC115956.1 | - | 7 | 1055 | 40056016 |
