siRNA Details
Gene Symbol: Fbxw24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013776.4 | NP_001013798.2 | 7 | 999 | ||
GenBank mRNA | BC080701.1 | - | 7 | 1000 | 30473126 | |
CO808277.1 | - | 2 | 347 | |||
DV651634.1 | - | 3 | 437 | |||
GQ418172.1 | - | 7 | 1000 |
