siRNA Details
Gene Symbol: Fbxw26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198674.2 | NP_941076.1 | 8 | 1189 | ||
GenBank mRNA | AK054298.1 | - | 8 | 1189 | ||
BC132114.1 | - | 8 | 1188 | 40130391 | ||
BC132304.1 | - | 8 | 1188 | 40130581 |
